missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Straight Air/Stream Inlet Tube
Introduce gases with precision. QuickFit™ straight air/steam inlet tube with ground glass cone upper end to connect with QuickFit™ sockets and lower end left plain for cutting to needed size.
Brand: Quickfit MF15/1
25.08 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed for introducing gases etc. into Quickfit™ assemblies
- Stem with integral ground glass cone to fit Quickfit™ sockets
- Lower end of tube is left plain for Glassblower to cut to length and draw to jet
Specifications
Air/Steam Inlet | |
7.4 mm | |
Designed for introducing gases | |
14/23 |
Borosilicate Glass | |
Quickfit™ assemblies | |
Straight |