Learn More
Quickfit™ Swan Neck Adapter
Has screwthread joint to accept thermometers and tubes
Brand: Quickfit MA6/33
63.60 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Has screwthread joint to accept thermometers and tubes
- Available in different socket, cone, and thread sizes
Specifications
24/29 | |
Borosilicate Glass | |
13 |
Ideal for securing thermometers, gas inlet tubes | |
Swan neck adapter |