Learn More
Quickfit™ Unbadged Joints Cones
Free from any ceramic marking
Brand:  Quickfit CNB29UB
 View more versions of this product
Product Code. 12614911
  • £73.95 / Pack of 10
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Free from any ceramic marking
  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Manufactured to accuracy which surpasses requirements of ISO 383
  • Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
  • Cones manufactured with shank length of 100mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Joint Cone
29/32
2mm