missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Vacuum Receiver Adapter

Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.

Brand:  Quickfit RA3/22/SC

62.39 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11382095

  • £119.00 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.

  • Delivery stem within cone
  • With sidearm for connection to a vacuum supply
  • Easy connection to flexible tubing via detachable plastic screwthread connector
TRUSTED_SUSTAINABILITY
Specifications

Specifications

19/26
5.51in.
Detachable Plastic screw thread connector, delivery stem, side-arm
Vacuum receiver angled adapter
Flexible tubing
140mm
Borosilicate Glass
19/26
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers