Learn More
Quickfit™ Vertical Splash Head Adapter
Usually inserted between a flask and condenser to prevent 'carry-over' of raw liquid
Brand:  Quickfit SH17/23
72.78 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11312115
  • £75.97 / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Usually inserted between a flask and condenser to prevent carry over of raw liquid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3)
24/29
Vertical Splash Head Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Vertical Splash Head Adapter > Cone size: 24/29; Socket size: 19/26