missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Quiltec™ Sterile Mop Head

Manufactured from 2-ply polyester around a rayon core. Contec™ Quiltec Sterile Mop Head is ideal for the highest grade of cleanroom due to the small numbers of particle and fibres.

Brand:  Contec™ GCQT0150

135.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.



Additional Details : Weight : 3.17000kg

 View more versions of this product

Product Code. 15995380

  • £164.00 / Pack of 40
Estimated Shipment: 22-11-2024
to see stock.
Add to basket

Description

Description

  • Two-ply 100% knitted polyester over a rayon core
  • Gamma irradiated at no less than 25 kGy
  • 5 mops double bagged
  • Approx 18-20sqm coverage
  • Certificates of conformity supplied with every batch
Specifications

Specifications

Mop Head
Not Autoclavable
Polyester
Sterile
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers