Learn More
Quinaldine, 95+%, Thermo Scientific Chemicals
£44.90 - £123.00
Chemical Identifiers
CAS | 91-63-4 |
---|---|
Molecular Formula | C10H9N |
Molecular Weight (g/mol) | 143.19 |
MDL Number | MFCD00006756 |
InChI Key | SMUQFGGVLNAIOZ-UHFFFAOYSA-N |
Synonym | quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech |
PubChem CID | 7060 |
IUPAC Name | 2-methylquinoline |
SMILES | CC1=CC=C2C=CC=CC2=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582911
|
Thermo Scientific Acros
132172500 |
250 mL | Glass bottle |
£44.90
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10085683
|
Thermo Scientific Acros
132170010 |
1 L | Glass bottle |
£123.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-63-4 | |
143.19 | |
SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
7060 | |
CC1=CC=C2C=CC=CC2=N1 |
C10H9N | |
MFCD00006756 | |
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
2-methylquinoline |
Specifications
91-63-4 | |
White to Yellow | |
248.0°C | |
Authentic | |
C10H9N | |
MFCD00006756 | |
15, 8163 | |
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
2-methylquinoline | |
7060 | |
95+% | |
Quinaldine, 97% |
-2.0°C | |
1.0580g/mL | |
79°C | |
95% min. (GC) | |
1.6070 to 1.6120 | |
20, 387 | |
1.058 | |
Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in chloroform,ether and alcohol | |
CC1=CC=C2C=CC=CC2=N1 | |
143.19 | |
143.19 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doct
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-085-1
missing translation for 'rtecsNumber' : UZ9625000
missing translation for 'tsca' : TSCA
Recommended Storage : Product may discolor to brownish on storage
RUO – Research Use Only