missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Quinidine, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Soluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 171°C to 173°C |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 91866 |
Sensitivity | Light sensitive |
Merck Index | 14,8060 |
Solubility Information | Soluble in methanol,chloroform,alcohol (1g/36ml),benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water. |
Formula Weight | 324.42 |
Chemical Name or Material | (+)-Quinidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'