missing translation for 'onlineSavingsMsg'
Learn More

(+)-Quinidine, Thermo Scientific Chemicals

Product Code. 11484254
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(+)-Quinidine is used to treat or prevent many types of irregular heartbeats (heart arrhythmias such as atrial fibrillation). It may also be used to treat malaria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol, chloroform, alcohol (1 g/36ml), benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00135581,MFCD00198096
InChI Key LOUPRKONTZGTKE-LHHVKLHASA-N
Synonym quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
PubChem CID 441074
ChEBI CHEBI:28593
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1

Specifications

Melting Point 171°C to 173°C
Quantity 5 g
UN Number UN2811
Beilstein 91866
Sensitivity Light sensitive
Merck Index 14,8060
Solubility Information Soluble in methanol,chloroform,alcohol (1g/36ml),benzene and DMSO(100mM). Slightly soluble in ether and petroleum ether. Insoluble in water.
Optical Rotation +260° (c=1 in Ethanol)
Formula Weight 324.42
Chemical Name or Material (+)-Quinidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'