missing translation for 'onlineSavingsMsg'
Learn More

Quinidine, Tocris Bioscience™

Product Code. 18732574
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Blocks sodium and potassium channels

Class IA antiarrythmic; reduces both Na+ and K+ channel currents, including INa, IKr and IKs. Prolongs QT and induces torsade de pointes (TdP).

Specifications

CAS 56-54-2
Molecular Formula C20H24N2O2
MDL Number MFCD00135581,MFCD00198096
Quantity 50 mg
Synonym quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
InChI Key LOUPRKONTZGTKE-LHHVKLHASA-N
SMILES COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Molecular Weight (g/mol) 324.42
PubChem CID 441074
ChEBI CHEBI:28593
Formula Weight 324.42
Percent Purity >98%
Chemical Name or Material Quinidine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'