missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinine hemisulfate monohydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
Solubility
Soluble in a mixture of chloroform and absolute alcohol (2:1).
Notes
Light sensitive. Incompatible with strong oxidizing agents, alkalis, ammonia, strong bases and iodine.
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
Solubility
Soluble in a mixture of chloroform and absolute alcohol (2:1).
Notes
Light sensitive. Incompatible with strong oxidizing agents, alkalis, ammonia, strong bases and iodine.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 217°C to 218°C (decomposition) |
Odor | Odorless |
Quantity | 10 g |
Beilstein | 6113937 |
Sensitivity | Light sensitive |
Merck Index | 14,8061 |
Solubility Information | Soluble in a mixture of chloroform and absolute alcohol (2:1). |
Optical Rotation | −241° (c=2 in 0.1M HCl) |
Formula Weight | 391.47 (373.46 Anhydrous) |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'