missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-boronic acid, tech. 90%, Thermo Scientific Chemicals

Product Code. 11395435
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-58-8
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.978
MDL Number MFCD01114698
InChI Key KXJJSKYICDAICD-UHFFFAOYSA-N
Synonym quinoline-8-boronic acid, 8-quinolineboronic acid, 8-quinolinylboronic acid, 8-quinoline boronic acid, quinolin-8-yl boronic acid, 8-boronoquinoline, 8-quinolylboronic acid, boronic acid, 8-quinolinyl, unii-5qs1a25ij6, quinolin-8-yl-8-boronic acid
PubChem CID 2734380
IUPAC Name quinolin-8-ylboronic acid
SMILES B(C1=C2C(=CC=C1)C=CC=N2)(O)O

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 147522
Merck Index 14,8069
Formula Weight 172.98
Percent Purity 99%
Chemical Name or Material Quinoline-8-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'