Learn More
Quinoline, 99%, Thermo Scientific Chemicals
£24.80 - £305.00
Chemical Identifiers
CAS | 91-22-5 |
---|---|
Molecular Formula | C9H7N |
Molecular Weight (g/mol) | 129.16 |
MDL Number | MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 |
InChI Key | SMWDFEZZVXVKRB-UHFFFAOYSA-N |
Synonym | chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol |
PubChem CID | 7047 |
ChEBI | CHEBI:17362 |
IUPAC Name | quinoline |
SMILES | C1=CC=C2N=CC=CC2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10032910
|
Thermo Scientific Acros
221140050 |
5 g | Glass bottle |
£26.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10032920
|
Thermo Scientific Acros
221141000 |
100 g | Glass bottle |
£24.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10419920
|
Thermo Scientific Acros
221145000 |
500 g | Glass bottle |
£83.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13428478
|
Thermo Scientific Acros
221140025 |
2.5 kg | Glass bottle |
£305.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
91-22-5 | |
129.16 | |
SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
7047 | |
quinoline |
C9H7N | |
MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
CHEBI:17362 | |
C1=CC=C2N=CC=CC2=C1 |
Specifications
91-22-5 | |
Gray to Purple | |
237.0°C | |
Authentic | |
C9H7N | |
MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987 | |
01,975; 02,356; 09,403 | |
1.095 | |
Solubility in water: slightly soluble. Other solubilities: soluble in hot water,miscible with alcohol,ether,acetone,benzene,and carbon disulfide | |
C1=CC=C2N=CC=CC2=C1 | |
129.16 | |
CHEBI:17362 | |
99% | |
Quinoline, 99% |
-15.0°C | |
1.0950g/mL | |
101°C | |
98.5% min. (GC) | |
1.6230 to 1.6260 | |
20,339 | |
15,8185 | |
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol | |
SMWDFEZZVXVKRB-UHFFFAOYSA-N | |
quinoline | |
7047 | |
129.16 | |
Crystalline Mass, Flakes, Powder and/or Chunks |
Safety and Handling
GHS H Statement
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful in contact with skin.
Toxic
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENT
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-051-6
missing translation for 'rtecsNumber' : VA9275000
missing translation for 'tsca' : TSCA
RUO – Research Use Only