missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinoline Yellow, Water Soluble 95%

Product Code. 10063400
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-92-0
Molecular Formula C18H9NNa2O8S2
Molecular Weight (g/mol) 477.37
MDL Number MFCD00080727
InChI Key FZUOVNMHEAPVBW-UHFFFAOYSA-L
PubChem CID 129893443
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Specifications

Assay Percent Range 95%
Solubility Information Solubility in water: 225g/L (20°C).
Infrared Spectrum Authentic
IUPAC Name disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
Percent Purity 95%
Grade Pure
Packaging Glass bottle
Quantity 25 g
Physical Form Powder
Chemical Name or Material Quinoline Yellow
compliance-icons
Product Identifier
  • Quinoline Yellow, water soluble
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'