missing translation for 'onlineSavingsMsg'
Learn More

Quinuclidine, 97+%, Thermo Scientific Chemicals

Product Code. 15400788
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.

Solubility
Soluble in alcohol, diethyl ether, water and organic solvents.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-76-5
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
MDL Number MFCD00006690
InChI Key SBYHFKPVCBCYGV-UHFFFAOYSA-N
Synonym quinuclidine, 1-azabicyclo 2.2.2 octane, 1,4-ethanopiperidine, 1,4-ethylenepiperidine, chinuclidin, abco, unii-xfx99fc5vi, 4-azabicyclo 2.2.2 octane, xfx99fc5vi
PubChem CID 7527
ChEBI CHEBI:38420
IUPAC Name 1-azabicyclo[2.2.2]octane
SMILES C1CN2CCC1CC2

Specifications

Melting Point 157°C to 160°C
Quantity 1 g
UN Number UN2811
Beilstein 103111
Sensitivity Air Sensitive
Merck Index 14,8081
Solubility Information Soluble in alcohol,diethyl ether,water and organic solvents.
Formula Weight 111.19
Percent Purity ≥97%
Chemical Name or Material Quinuclidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'