missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinuclidine, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.
Solubility
Soluble in alcohol, diethyl ether, water and organic solvents.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.
Solubility
Soluble in alcohol, diethyl ether, water and organic solvents.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Specifications
Specifications
Melting Point | 157°C to 160°C |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 103111 |
Sensitivity | Air Sensitive |
Merck Index | 14,8081 |
Solubility Information | Soluble in alcohol,diethyl ether,water and organic solvents. |
Formula Weight | 111.19 |
Percent Purity | ≥97% |
Chemical Name or Material | Quinuclidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'