missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-(3-Chlorophenyl)ethylamine, ChiPros™, 99%, ee 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Boiling Point | 112°C (13 mmHg) |
Quantity | 1 g |
UN Number | UN2735 |
Solubility Information | Insoluble in water. |
Formula Weight | 155.63 |
Percent Purity | 99% |
Assay | ee ≥98% |
Chemical Name or Material | (R)-1-(3-Chlorophenyl)ethylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'