missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Cyclohexylethyl isocyanate, 94%, Thermo Scientific™

Product Code. 15449139
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93470-26-9
Boiling Point 210°C to 215°C
Molecular Formula C9H15NO
Refractive Index 1.465
MDL Number MFCD05664073
Quantity 1 g
UN Number UN2206
Beilstein 9255848
Sensitivity Moisture sensitive
Synonym r---1-cyclohexylethyl isocyanate, r-1-cyclohexylethylisocyanate, 1r-1-isocyanatoethyl cyclohexane, r-1-cyclohexylethyl isocyanate, 1-isocyanatoethylcyclohexane
InChI Key IGSKYRAPJLTXSO-MRVPVSSYSA-N
SMILES CC(C1CCCCC1)N=C=O
IUPAC Name [(1R)-1-isocyanatoethyl]cyclohexane
Molecular Weight (g/mol) 153.225
PubChem CID 51387275
Formula Weight 153.23
Percent Purity 94%
Chemical Name or Material (R)-(-)-1-Cyclohexylethyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'