missing translation for 'onlineSavingsMsg'
Learn More

BUCHI R-100 System

Entry level Rotavapor to meet the essential needs in evaporation and BUCHI’s high quality standards.

Brand:  BUCHI 11100V1011

 View more versions of this product

Product Code. 15344906

This item is not returnable. View return policy

Description

Description

  • R-100 rotary evaporator
  • Entry level Rotavapor
  • Meets the essential needs in evaporation and BUCHI’s high quality standards.
Specifications

Specifications

No Protective coating
5 L
Ambient to 95°C
Vertical
220/240 V
280 rpm
F-100, fix 10°C
SJ 29/32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BUCHI R-100 System > F-100, fix 10 °C

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.