missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

Product Code. 15405475
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-84-5
Molecular Formula C48H28F24O4P2
Molecular Weight (g/mol) 1186.66
MDL Number MFCD19443625
InChI Key JDKXXRZDVSJSNQ-UHFFFAOYSA-N
Synonym r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxybiphenyl, ar-2,2',4,4'-tetramethoxy-6,6'-bis bis 3,5-bis trifluoromethyl phenyl phosphino biphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, r-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis bis 3,5-bis trifluoromethyl phenyl phosphine, s-2,2'-bis bis 3,5-trifluoromethylphenyl phosphino-4,4',6,6'-tetramethoxy-1,1'-biphenyl, bis 3,5-bis trifluoromethyl phenyl 2'-bis 3,5-bis trifluoromethyl phenyl phosphanyl-4,4',6,6'-tetramethoxy-1,1'-biphenyl-2-yl phosphane
PubChem CID 87111592
IUPAC Name [2-[2-bis[3,5-bis(trifluoromethyl)phenyl]phosphanyl-4,6-dimethoxyphenyl]-3,5-dimethoxyphenyl]-bis[3,5-bis(trifluoromethyl)phenyl]phosphane
SMILES COC1=CC(OC)=C(C(=C1)P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=C(OC)C=C(OC)C=C1P(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 1186.64
Percent Purity ≥97%
Chemical Name or Material (R)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'