missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2-Amino-3-methylbutane, ChiPros 98%, ee 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
(R)-(-)-2-Amino-3-methylbutane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water (60g/L at 20°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under dry inert gas. Incompatible with oxidizing agents. Store in cool place. It is sensitive to air.
(R)-(-)-2-Amino-3-methylbutane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water (60g/L at 20°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under dry inert gas. Incompatible with oxidizing agents. Store in cool place. It is sensitive to air.
Specifications
Specifications
Density | 0.746 |
Boiling Point | 85°C to 87°C |
Flash Point | <21°C (69°F) |
Refractive Index | 1.406 |
Quantity | 5 g |
UN Number | UN2733 |
Beilstein | 1718798 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water (60g/L at 20°C). |
Optical Rotation | −5.2° (Neat) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'