missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminononane, ChiPros 99+%, ee 98+%, Thermo Scientific Chemicals

Product Code. 11304226
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74069-74-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.274
MDL Number MFCD03844739
InChI Key ALXIFCUEJWCQQL-SECBINFHSA-N
Synonym r-2-aminononane, r---2-aminononane, 2r-nonan-2-amine, r-2-nonanamine, r-2-nonylamine, 2-nonanamine, 2r, r-2-aminononane, chipros r , produced by basf
PubChem CID 22831496
IUPAC Name (2R)-nonan-2-amine
SMILES CCCCCCCC(C)N

Specifications

Density 0.782
Boiling Point 73°C (19 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.4271
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Optical Rotation −6° (Neat)
Formula Weight 143.27
Percent Purity ≥99%
Assay ee ≥98%
Chemical Name or Material (R)-(-)-2-Aminononane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'