missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals

Product Code. 11935431
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397246-14-9
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
InChI Key KLRRFBSWOIUAHZ-SSDOTTSWSA-N
Synonym r-+-2-boc-amino-1,4-butanediol, r-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, r-2-boc-amino-butane-1,4-diol, r-2-boc-amino-1,4-butanediol, tert-butyl n-2r-1,4-dihydroxybutan-2-yl carbamate, pubchem15259, ksc498c9f, 1r-3-hydroxy-1-hydroxymethyl propyl carbamic acid tert-butyl ester, r-2-boc-amino-butan-1,4-diol, 2r-tert-butoxycarbonylamino butane-1,4-diol
PubChem CID 11954482
IUPAC Name tert-butyl N-[(2R)-1,4-dihydroxybutan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CCO)CO

Specifications

Melting Point 66.4°C to 72.1°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Specific Rotation 7.8
Quantity 1 g
Formula Weight 205.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-2-(BOC-Amino)-1, 4-butanediol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-2-(BOC-Amino)-1,4-butanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'