missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-(BOC-amino)-1-propanol, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | 56°C to 60°C |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (GC) |
Linear Formula | HOCH2CH(CH3)NNCO2C(CH3)3 |
Packaging | Glass Bottle |
Formula Weight | 175.23 |
Percent Purity | 98% |
Quantity | 1 g |
Chemical Name or Material | (R)-2-(BOC-Amino)-1-propanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'