missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-(Boc-amino)butyric acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
Solubility
Soluble in DMF (1mmol in 1ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
Solubility
Soluble in DMF (1mmol in 1ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications
Specifications
Color | Yellow |
Quantity | 1 g |
Solubility Information | Soluble in DMF (1mmol in 1ml DMF). |
Formula Weight | 203.24 |
Percent Purity | 95% |
Physical Form | Liquid |
Chemical Name or Material | (R)-2-(Boc-amino)butyric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'