missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals

Product Code. 10451484
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112022-83-0, 108-88-3
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078440
InChI Key VMKAFJQFKBASMU-QGZVFWFLSA-N
Synonym r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
PubChem CID 9838490
IUPAC Name (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 0.9500g/mL
Boiling Point 111.0°C
Flash Point 4°C
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 0.95
Solubility Information Solubility in water: reacts
Formula Weight 277.17
Chemical Name or Material (R)-(+)-2-Methyl-CBS-oxazaborolidine
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.