missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methylpiperazine, 98+%, Thermo Scientific Chemicals

Product Code. 11426106
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75336-86-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00192317
InChI Key JOMNTHCQHJPVAZ-RXMQYKEDSA-N
Synonym r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
PubChem CID 7330434
IUPAC Name (2R)-2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 76°C to 78°C
Boiling Point 155°C to 156°C
Flash Point 65°C (149°F)
Quantity 1 g
UN Number UN2925
Beilstein 3647994
Sensitivity Air sensitive; Hygroscopic
Optical Rotation −7° (c=1 in Ethanol)
Formula Weight 100.17
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-Methylpiperazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'