Learn More
(R)-(-)-2-Methylpyrrolidine, 95%, Thermo Scientific Chemicals
£34.30 - £97.00
Chemical Identifiers
CAS | 41720-98-3 |
---|---|
Molecular Formula | C5H11N |
Molecular Weight (g/mol) | 85.15 |
MDL Number | MFCD07783026 |
InChI Key | RGHPCLZJAFCTIK-RXMQYKEDSA-N |
Synonym | r-2-methylpyrrolidine, r-2-methyl-pyrrolidine, 2r-2-methylpyrrolidine, r---2-methylpyrrolidine, r-2-methyl pyrrolidine, 2-r-methylpyrrolidine, 2r---2-methyltetrahydropyrrole, 2r---2-methylpyrrolidine, pyrrolidine, 2-methyl-, 2r, 2-methyl-pyrrolidine |
PubChem CID | 641544 |
ChEBI | CHEBI:78858 |
IUPAC Name | (2R)-2-methylpyrrolidine |
SMILES | CC1CCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15464278
|
Thermo Scientific Alfa Aesar
H57885.MD |
250 mg |
£34.30
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15454278
|
Thermo Scientific Alfa Aesar
H57885.03 |
1 g |
£97.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a building block used in a multikilogram synthesis of a chiral naphthalenoid histamine receptor antagonist.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Keep container in a cool refrigerated, well-ventilated area. Moisture sensitive
Chemical Identifiers
41720-98-3 | |
85.15 | |
RGHPCLZJAFCTIK-RXMQYKEDSA-N | |
641544 | |
(2R)-2-methylpyrrolidine |
C5H11N | |
MFCD07783026 | |
r-2-methylpyrrolidine, r-2-methyl-pyrrolidine, 2r-2-methylpyrrolidine, r---2-methylpyrrolidine, r-2-methyl pyrrolidine, 2-r-methylpyrrolidine, 2r---2-methyltetrahydropyrrole, 2r---2-methylpyrrolidine, pyrrolidine, 2-methyl-, 2r, 2-methyl-pyrrolidine | |
CHEBI:78858 | |
CC1CCCN1 |
Specifications
41720-98-3 | |
7°C (45°F) | |
1.43 | |
UN2733 | |
Insoluble in water. | |
−19° (c=5 in Methanol) | |
(2R)-2-methylpyrrolidine | |
641544 | |
85.15 | |
(R)-(-)-2-Methylpyrrolidine |
0.84 | |
C5H11N | |
MFCD07783026 | |
r-2-methylpyrrolidine, r-2-methyl-pyrrolidine, 2r-2-methylpyrrolidine, r---2-methylpyrrolidine, r-2-methyl pyrrolidine, 2-r-methylpyrrolidine, 2r---2-methyltetrahydropyrrole, 2r---2-methylpyrrolidine, pyrrolidine, 2-methyl-, 2r, 2-methyl-pyrrolidine | |
RGHPCLZJAFCTIK-RXMQYKEDSA-N | |
CC1CCCN1 | |
85.15 | |
CHEBI:78858 | |
95% |
Safety and Handling
GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only