missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing /reducing agents, strong acids, strong bases.
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing /reducing agents, strong acids, strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 158°C to 160°C |
Quantity | 5 g |
Beilstein | 82133 |
Solubility Information | Soluble in water. |
Optical Rotation | +10.5° (c=5 in Water) |
Formula Weight | 129.12 |
Percent Purity | ≥98% |
Chemical Name or Material | (R)-(+)-2-Pyrrolidinone-5-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'