Learn More
(R)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific™
£136.00 - £201.40
Chemical Identifiers
CAS | 170564-98-4 |
---|---|
MDL Number | MFCD01311791 |
InChI Key | SEQXIQNPMQTBGN-SECBINFHSA-N |
Synonym | r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853 |
PubChem CID | 7016858 |
IUPAC Name | (3R)-3-amino-3-phenylpropan-1-ol |
SMILES | C1=CC=C(C=C1)C(CCO)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10272712
|
Thermo Scientific Acros
363125000 |
500 mg | Glass bottle | ||||||
10180802
|
Thermo Scientific Acros
363120010 |
1 g | Glass bottle |
£136.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
170564-98-4 | |
SEQXIQNPMQTBGN-SECBINFHSA-N | |
7016858 | |
C1=CC=C(C=C1)C(CCO)N |
MFCD01311791 | |
r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853 | |
(3R)-3-amino-3-phenylpropan-1-ol |
Specifications
170564-98-4 | |
100.0 | |
97.5% min. | |
95% | |
MFCD01311791 | |
SEQXIQNPMQTBGN-SECBINFHSA-N | |
(3R)-3-amino-3-phenylpropan-1-ol | |
7016858 | |
≥94% | |
(R)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.) |
94.0 | |
White to Yellow or Yellow | |
Authentic | |
C9H13NO | |
r-1-phenyl-3-propanolamine, r-3-amino-3-phenylpropan-1-ol, 3r-3-amino-3-phenylpropan-1-ol, r-beta-phenylalaninol, r-3-amino-3-phenyl-1-propanol, r-3-phenyl-beta-alaninol, r-3-amino-3-phenyl-propan-1-ol, r--phenylalaninol, pubchem13853 | |
C1=CC=C(C=C1)C(CCO)N | |
151.209 | |
151.21 | |
Viscous Liquid or Low Melting Solid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'tsca' : TSCA
- (R)-3-Amino-3-phenylpropan-1-ol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only