missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)piperidine, 97%

Product Code. 15459487
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15459487

Brand: Thermo Scientific Alfa Aesar H52749.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 309956-78-3 | C10H20N2O2 | 200.282 g/mol

(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 309956-78-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03093382
InChI Key WUOQXNWMYLFAHT-MRVPVSSYSA-N
Synonym r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester
PubChem CID 1514172
IUPAC Name tert-butyl N-[(3R)-piperidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Solubility Information Soluble in methanol and ethanol.
Quantity 1 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)piperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.