missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Chloro-1,2-propanediol, 97%, ee 98%, Thermo Scientific Chemicals

Product Code. 11319405
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57090-45-6
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00135169
InChI Key SSZWWUDQMAHNAQ-VKHMYHEASA-N
Synonym r---3-chloro-1,2-propanediol, r-3-chloro-1,2-propanediol, 2r-3-chloropropane-1,2-diol, r-3-chloropropane-1,2-diol, alpha-chlorohydrin,-, unii-64ymo02kae, ccris 7387, 1,2-propanediol, 3-chloro-, r, 64ymo02kae, 1,2-propanediol, 3-chloro-, 2r
PubChem CID 148793
ChEBI CHEBI:18663
IUPAC Name (2R)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O

Specifications

Density 1.322
Boiling Point 213°C
Flash Point 137°C (279°F)
Refractive Index 1.48
Quantity 1 g
UN Number UN2689
Beilstein 2202928
Optical Rotation −0.9° (Neat)
Formula Weight 110.54
Percent Purity 97%
Assay ee 98%
Chemical Name or Material (R)-(-)-3-Chloro-1,2-propanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'