missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Pyrrolidinol hydrochloride, 98%, Thermo Scientific™
£23.40 - £160.27
Chemical Identifiers
CAS | 104706-47-0 |
---|---|
Molecular Formula | C4H9NO·ClH |
Molecular Weight (g/mol) | 123.58 |
InChI Key | QPMSJEFZULFYTB-PGMHMLKASA-N |
Synonym | r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride |
PubChem CID | 2759336 |
IUPAC Name | (3R)-pyrrolidin-3-ol;hydrochloride |
SMILES | C1CNCC1O.Cl |
Description
Chemical Identifiers
104706-47-0 | |
123.58 | |
r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride | |
(3R)-pyrrolidin-3-ol;hydrochloride |
C4H9NO·ClH | |
QPMSJEFZULFYTB-PGMHMLKASA-N | |
2759336 | |
C1CNCC1O.Cl |
Specifications
104706-47-0 | |
97.5% min. (Argentometry) | |
C4H9NO·ClH | |
r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride | |
C1CNCC1O.Cl | |
123.58 | |
123.58 | |
(R)-3-Pyrrolidinol hydrochloride |
102°C to 107.5°C | |
Glass Bottle | |
-7.6 | |
QPMSJEFZULFYTB-PGMHMLKASA-N | |
(3R)-pyrrolidin-3-ol;hydrochloride | |
2759336 | |
98% |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (R)-3-Pyrrolidinol hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only