missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Methylmandelic acid, ChiPros, 98%, ee 97+%, Thermo Scientific Chemicals

Product Code. 11425730
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31284-89-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00798433,MFCD00961860
InChI Key SFGURAWGCAPHON-UHFFFAOYNA-N
Synonym 2-hydroxy-2-4-methylphenyl acetic acid, 4-methylmandelic acid, p-methyl mandelic acid, 2-hydroxy-2-p-tolyl acetic acid, hydroxy 4-methylphenyl acetic acid, hydroxy-p-tolyl-acetic acid, p-methylmandelic acid, 4-methylmaldelic acid, p-methyl-mandelic acid, acmc-1afsj
PubChem CID 12458823
IUPAC Name (2R)-2-hydroxy-2-(4-methylphenyl)acetic acid
SMILES CC1=CC=C(C=C1)C(O)C(O)=O

Specifications

Melting Point 130°C to 133°C
Quantity 25 g
Sensitivity Light sensitive
Optical Rotation −154° (c=1 in Methanol)
Formula Weight 166.18
Percent Purity 98%
Assay ee ≥97%
Chemical Name or Material (R)-(-)-4-Methylmandelic acid, ChiPros

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'