Learn More
IKA™ R 50 Stirring Shaft, High Speed
For rapid mixing, dissolving, and disagglomerating pigment agglomerates.
Brand:  IKA™ 0001689300
Product Code. 10508361
  • £2377.00 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Turns the T 50 digital stirrer into a high speed stirrer.
  • For operational safety, a protective cage is fitted around the mixing element.
  • Stainless steel construction.
Specifications
Specifications
0.25 to 30 L
T 50 digital Ultra-Turrax stirrer
Stainless Steel (316)
Immersion depth: 180 mm
Stirring Shaft