missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ R 568 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Positive allosteric modulator and allosteric agonist of the human calcium-sensing receptor (hCaSR). Exhibits calcimimetic activity. Has the ability to restore function to hCaSR mutants that cause familial hypocalciuric hypercalcemia (FHH) and neonatal severe hyperparathyroidism (NSHPT). Exerts a suppressive effect on parathyroid (PT) hormone secretion; inhibits PT cell proliferation in rats with renal insufficiency.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 340.29 |
| Purity | >99% |
| Molecular Weight (g/mol) | 340.288 |
| Chemical Name or Material | R 568 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction