missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-8-Hydroxy-DPAT hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Full 5-HT1A serotonin receptor agonist. Reduces hippocampal 5-HT levels following systemic administration in rats in vivo. More active enantiomer of 8-Hydroxy-DPAT hydrobromide .
Racemate and 7-Hydroxy Isomer also available.
Specifications
Quantity | 10 mg |
Formula Weight | 328.29 |
Percent Purity | >99% |
Chemical Name or Material | (R)-(+)-8-Hydroxy-DPAT hydrobromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'