missing translation for 'onlineSavingsMsg'
Learn More

(R)-CPP, Tocris Bioscience™

Product Code. 18074432
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent NMDA antagonist; more active enantiomer of (RS)-CPP

Highly potent NMDA antagonist; more active isomer. Shows some selectivity for GluN2A (formally NR2A) containing receptors (Ki values are 0.041, 0.27, 0.63 and 1.99 μM for inhibition of GluN2A-, GluN2B-, GluN2C- and GluN2D-containing recombinant NMDA receptors respectively).

Racemate also available.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 126453-07-4
Molecular Formula C8H17N2O5P
Molecular Weight (g/mol) 252.207
InChI Key CUVGUPIVTLGRGI-SSDOTTSWSA-N
Synonym r-cpp, chembl47277, 2r-4-3-phosphonopropyl piperazine-2-carboxylic acid, 3-r-2-carboxypiperazin-4-yl-propyl-1-phosphonic acid, tocris-0173, tocris-0247, lopac-c-104, 2-piperazinecarboxylicacid, 4-3-phosphonopropyl-, 2r, --cpp, r-4-3-phosphonopropyl piperazine-2-carboxylic acid
PubChem CID 6603754
IUPAC Name (2R)-4-(3-phosphonopropyl)piperazine-2-carboxylic acid
SMILES C1CN(CC(N1)C(=O)O)CCCP(=O)(O)O

Specifications

Quantity 50 mg
Formula Weight 252.21
Percent Purity >98%
Chemical Name or Material (R)-CPP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'