Learn More
R&D Systems™ Ac-Arg-Gly-Lys(Ac)-AMC Fluorogenic Peptide Substrate
Substrate for certain deacetylases such as histone deacetylases
Brand:  R&D Systems™ ES016
196.97 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18108320
  • £259.00 / 5mg
Explore more special offers
Description
Description
  • Hydrolysis of Lys-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • After the enzymatic removal of the acetyl group, the substrate can be cleaved by trypsin-like enzymes
  • Purity >95%
Specifications
Specifications
Ac-Arg-Gly-Lys(Ac)-AMC (Ac-RGK(Ac)-AMC)
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.