missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bovine Fibronectin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£184.66 - £384.44
Specifications
Formulation | Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea. |
---|---|
Gene ID (Entrez) | 280794 |
Name | Fibronectin |
Source | Bovine plasma-derived Fibronectin protein |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. Avoid vortexing and excessive agitation. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18399237
|
R&D Systems™
1030-FN-05M |
5mg |
£384.44
5mg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18370568
|
R&D Systems™
1030-FN-01M |
1mg |
£184.66
1mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Bovine Fibronectin Protein is derived from Bovine Plasma. The Bovine Fibronectin Protein has been validated for the following applications: Bioactivity.Specifications
Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea. | |
Fibronectin | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. Avoid vortexing and excessive agitation. | |
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
280794 | |
Bovine plasma-derived Fibronectin protein | |
Unconjugated |