missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex 3-D Culture Matrix RGF Basement Membrane Extract
Applications: Bioactivity
£86.39 - £475.15
Specifications
Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |
---|---|
Concentration | 14 to 16 mg/mL |
Endotoxin Level | ≤8EU/mL |
For Use With (Application) | Bioactivity |
Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18481270
|
R&D Systems™
3445-001-01 |
1 mL |
£86.39
1mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18462400
|
R&D Systems™
3445-010-01 |
2 x 5 mL |
£475.15
10mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18462020
|
R&D Systems™
3445-005-01 |
5 mL |
£278.61
5mL |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Functional assays: Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. 3D Culture - Basement Membrane Extract promotes differentiation of a human epithelial cell line derived from mammary gland (MCF-10A) or human prostate (PC-3) into acinar structures. Gelling assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.Specifications
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. | |
≤8EU/mL | |
Murine Engelbreth-Holm-Swarm (EHS) tumor. |
14 to 16 mg/mL | |
Bioactivity |