missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Basement Membrane Extract, Pathclear (w/ Phenol Red)

soluble form of basement membrane purified from Engelbreth-Holm-Swarm (EHS) tumor

£219.00 - £348.00

Specifications

Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
Concentration 12 to 18 mg/mL
Endotoxin Level ≤8EU/mL
For Use With (Application) Bioactivity
Source Murine Engelbreth-Holm-Swarm (EHS) tumor.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18401180
View Documents Promotion Details
R&D Systems™
3432-005-01P
5 mL
£219.00
5mL
Please to purchase this item. Need a web account? Register with us today!
18452390
View Documents
R&D Systems™
3432-010-01P
2 x 5 mL
£348.00
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The extract gels at 37°C to form a reconstituted basement membrane. The major components of BME include laminin, collagen IV, entactin, and heparin sulfate proteoglycan. BME can be used in a multiple applications, under a variety of cell culture conditions, for maintaining growth or promoting differentiation of primary endothelial, epithelial, smooth muscle and stem cells. BME can also be utilized in cell attachment, neurite outgrowth, angiogenesis, in vitro cell invasion and in vivo tumorigenicity assays.
Specifications

Specifications

Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
≤8EU/mL
Murine Engelbreth-Holm-Swarm (EHS) tumor.
12 to 18 mg/mL
Bioactivity
Videos
SDS
Documents

Documents

Product Certifications