Learn More
Tocris Bioscience™ EPZ 004777
Highly potent DOT1L inhibitor
Brand: Tocris Bioscience™ 5567/10
Additional Details : CAS Number : 1338466-77-5 Weight : 0.00970kg
Description
EPZ 004777 is a highly potent DOT1L inhibitor (IC50 = 0.4 nM). Exhibits >1000-fold selectivity for DOT1L over a panel of other methyltransferases. Selectively inhibits proliferation and induces apoptosis of MLL-rearranged cells in vitro. Prolongs survival in a MLL xenograft mouse model. Enhances efficiency of Yamanaka-factor factor reprogramming of fibroblasts by 3-4-fold.Specifications
EPZ 004777 | |
7-[5-Deoxy-5-[[3-[[[[4-(1,1-dimethylethyl)phenyl]amino]carbonyl]amino]propyl](1-methylethyl)amino]-β-D-ribofuranosyl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine | |
Soluble to 100mM in DMSO and to 100mM in ethanol | |
539.67 | |
Bioactive Small Molecule |
1338466-77-5 | |
10 mg | |
C28H41N7O4 | |
Solid | |
>98% |
Research Use Only. Not For Human or Veterinary Use