missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flow Cytometry Mouse Lyse Buffer (10X), R&D Systems™
Applications: Flow Cytometry
Brand: R&D Systems FC003
Additional Details : Weight : 0.09700kg
Description
- Gently lyses mouse erythrocytes
Applications:
Flow Cytometry
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. | |
Lysis Buffer | |
50mL |
Upto 6 Months | |
Spleen or whole blood samples |