missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£204.10
Specifications
Gene ID (Entrez) | 650 |
---|---|
Molecular Weight (g/mol) | 12.8 kDa (monomer) |
Name | BMP-2 |
Quantity | 10μg |
Source | E. coli-derived BMP-2 protein Ala284-Arg396 This form is not detected using human BMP-2 Quantikine ELISA (Catalog &Num; DBP200 ). |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18158280
|
R&D Systems™
355-BEC-010 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£204.10
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18324967
|
R&D Systems™
355-BEC-010/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£204.10
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed) is derived from E. coli. The Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed) has been validated for the following applications: Bioactivity.Specifications
650 | |
BMP-2 | |
E. coli-derived BMP-2 protein Ala284-Arg396 This form is not detected using human BMP-2 Quantikine ELISA (Catalog &Num; DBP200 ). | |
Disulfide-linked homodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
12.8 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC | |
Recombinant |