Learn More
R&D Systems™ Human C1qR1/CD93 (NP_036204) VersaClone cDNA
Complement Component C1q Receptor
Brand:  R&D Systems™ RDC0590
Product Code. 18452190
  • £648.00 / 10µg
Explore more special offers
Description
Description
The Human C1q R1/CD93 (NP_036204) VersaClone cDNA clone contains the complete ORF for C1qR1/CD93, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The C1qR1/CD93 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hCD93
10 μg at 0.2 μg/μL
1972bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
NP_036204
cDNA is provided in 10mM Tris-Cl, pH 8.5
AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans