missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Chitotriosidase/CHIT1 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 3559-GH-010
This item is not returnable.
View return policy
Description
Measured by its ability to cleave 4-Methylumbelliferyl-beta-D-N,N’,N"e;-triacetylchitotriose (4-MU-TACT). The specific activity is >1,500 pmol/min/μg, as measured under the described conditions.
Specifications
NP_003456 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 52 kDa, reducing conditions; M.W. (theoretical): 51 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
CHIT1 | |
Recombinant | |
Chitotriosidase/CHIT1 |
Enzyme Activity | |
1118 | |
Chitotriosidase/CHIT1 | |
Mouse myeloma cell line, NS0-derived human Chitotriosidase/CHIT1 protein Ala22-Asn466, with a C-terminal 10-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human Chitotriosidase/CHIT1 Protein, CF > 10ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction