Learn More
R&D Systems™ Human Coagulation Factor II/Thrombin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 2196-SE-200
Product Code. 18328947
  • £173.00 / 200µg
Explore more special offers
Description
Description
The Human Coagulation Factor II/Thrombin Protein is derived from Human Plasma. The Human Coagulation Factor II/Thrombin Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in MES, NaCl and Brij-35.
Coagulation Factor II/Thrombin
Human plasma-derived Coagulation Factor II/Thrombin protein The human plasma used for the isolation of this product was certified by the supplier to be non-reactive for HIV-1/2 and HBsAg negative at the donor level. Human blood products should always be treated in accordance with universal handling precautions.
pro form
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
2147
200 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human Coagulation Factor II/Thrombin Protein > 200ug; Carrier Free