missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CXCR2/IL-8RB (AAB25880) VersaClone cDNA
Interleukin 8 Receptor B
Brand: R&D Systems™ RDC0027
Additional Details : Weight : 0.00970kg
Description
The Human CXCR2/IL-8 RB (AAB25880) VersaClone cDNA clone contains the complete ORF for CXCR2/IL-8RB, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CXCR2/IL-8RB insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hCXCR2 | |
10 μg at 0.2 μg/μL | |
1081bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
AAB25880 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8 R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
Human |
For Reasearch Use Only; Not for Use in Humans