missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human FATP2 (AAC64973) VersaClone cDNA
Fatty Acid Transport Protein 2
Brand: R&D Systems™ RDC0477
Additional Details : Weight : 0.00970kg
Description
The Human FATP2 (AAC64973) VersaClone cDNA clone contains the complete ORF for FATP2/SLC27A2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FATP2/SLC27A2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hFATP2 | |
10 μg at 0.2 μg/μL | |
1876bp | |
Human |
AAC64973 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans